Assay Details
Gene Symbol: SLA-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK232210.1 | - | 2 - 3 | 495 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Gene Symbol: SLA-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK232210.1 | - | 2 - 3 | 495 | 64 |
Set Membership: |
![]() ![]() ![]() ![]() |