This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06919264_m1
See other TIRAP GE Assays ›
Gene Symbol:
TIRAP
Gene Name:
TIR domain containing adaptor protein
Gene Aliases:
-
Chromosome Location:
Chr.9: 53374559 - 53394172 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06919264_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TIRAP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003130060.5 XP_003130108.1 2019 77
XM_005667493.3 XP_005667550.1 884 77
XM_021063110.1 XP_020918769.1 3237 77
XM_021063111.1 XP_020918770.1 1890 77
XM_021063112.1 XP_020918771.1 5092 77
XM_021063114.1 XP_020918773.1 2296 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0