This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06918267_m1
See other TOGARAM1 GE Assays ›
Gene Symbol:
TOGARAM1
Gene Name:
TOG array regulator of axonemal microtubules 1
Gene Aliases:
FAM179B
Chromosome Location:
Chr.1: 175198172 - 175274309 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06918267_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TOGARAM1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005659920.3 XP_005659977.1 5034 148
XM_005659921.3 XP_005659978.1 5016 148
XM_005659923.3 XP_005659980.1 4935 148
XM_005659924.3 XP_005659981.1 4881 148
XM_013993373.2 XP_013848827.1 4863 148
XM_013993374.2 XP_013848828.1 4782 148
XM_013993375.2 XP_013848829.1 4552 148
XM_021101137.1 XP_020956796.1 4917 148
XM_021101162.1 XP_020956821.1 4764 148
XM_021101164.1 XP_020956823.1 4534 148

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Sus scrofa

Panther Classification:

Molecular Function -

non-motor microtubule binding protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0