Assay Details
Gene Symbol: MAGOH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003127999.5 | XP_003128047.2 | 636 | 90 | ||
GenBank mRNA | AK396533.1 | - | 2 - 3 | 695 | 90 | |
AK399749.1 | - | 2 - 3 | 226 | 90 | ||
AY610175.2 | - | 2 - 3 | 180 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
