This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06912460_m1
See other NHSL1 GE Assays ›
Gene Symbol:
NHSL1
Gene Name:
NHS like 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 25663554 - 25988106 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06912460_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NHSL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005659173.3 XP_005659230.1 6426 103
XM_005659175.3 XP_005659232.1 6426 103
XM_005659176.3 XP_005659233.1 2601 103
XM_021087394.1 XP_020943053.1 2967 103
XM_021087398.1 XP_020943057.1 2601 103
XM_021087404.1 XP_020943063.1 2967 103
XM_021087406.1 XP_020943065.1 442 103
XM_021087407.1 XP_020943066.1 409 103
GenBank mRNA AK395859.1 - 3 - 4 1134 103

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_021087406.1, XM_021087407.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0