This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06911250_m1
See other SGMS2 GE Assays ›
Gene Symbol:
SGMS2
Gene Name:
sphingomyelin synthase 2
Gene Aliases:
-
Chromosome Location:
Chr.8: 114052726 - 114145946 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06911250_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SGMS2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_013978971.2 XP_013834425.1 1163 95
XM_013978972.2 XP_013834426.1 1213 95
XM_013978974.2 XP_013834428.1 997 95
XM_013978976.2 XP_013834430.1 941 95
XM_013978980.2 XP_013834434.1 1104 95
XM_021101689.1 XP_020957348.1 1167 95
XM_021101691.1 XP_020957350.1 1001 95
XM_021101692.1 XP_020957351.1 1097 95
GenBank mRNA AK394800.1 - 3 - 4 929 95

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0