This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06911158_m1
See other TUT4 GE Assays ›
Gene Symbol:
TUT4
Gene Name:
terminal uridylyl transferase 4
Gene Aliases:
ZCCHC11
Chromosome Location:
Chr.6: 159655658 - 159794450 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06911158_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TUT4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021096634.1 XP_020952293.1 4485 72
XM_021096635.1 XP_020952294.1 4482 72
XM_021096636.1 XP_020952295.1 4470 72
XM_021096637.1 XP_020952296.1 4485 72
XM_021096638.1 XP_020952297.1 4482 72
XM_021096639.1 XP_020952298.1 4470 72
XM_021096640.1 XP_020952299.1 4272 72
XM_021096641.1 XP_020952300.1 4272 72
XM_021096642.1 XP_020952301.1 4269 72
XM_021096643.1 XP_020952302.1 4257 72
XM_021096644.1 XP_020952303.1 4152 72
XM_021096645.1 XP_020952304.1 4137 72
GenBank mRNA AK394735.1 - 8 - 9 889 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

nucleotidyltransferase transferase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0