This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06907109_m1
See other DMAC2L GE Assays ›
Gene Symbol:
DMAC2L
Gene Name:
distal membrane arm assembly complex 2 like
Gene Aliases:
ATP5S
Chromosome Location:
Chr.1: 180069157 - 180081332 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06907109_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DMAC2L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001926336.6 XP_001926371.3 697 134
XM_013993389.2 XP_013848843.1 637 134
XM_021101490.1 XP_020957149.1 694 134
XM_021101506.1 XP_020957165.1 634 134
GenBank mRNA AK391697.1 - 4 - 5 587 134

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0