This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06905510_m1
See other PRRT2 GE Assays ›
Gene Symbol:
PRRT2
Gene Name:
proline rich transmembrane protein 2
Gene Aliases:
-
Chromosome Location:
Chr.3: 18048614 - 18052481 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06905510_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRRT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021086426.1 XP_020942085.1 184 74
XM_021086427.1 XP_020942086.1 180 74
XM_021086428.1 XP_020942087.1 205 74
GenBank mRNA AK390546.1 - 1 - 2 112 74

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_021086426.1, XM_021086427.1, XM_021086428.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 3' Most Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0