This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06904747_g1
See other BET1L GE Assays ›
Gene Symbol:
BET1L
Gene Name:
Bet1 golgi vesicular membrane trafficking protein like
Gene Aliases:
-
Chromosome Location:
Chr.2: 26475 - 31960 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06904747_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BET1L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003124224.4 XP_003124272.1 263 63
XM_003354407.4 XP_003354455.1 260 63
XM_013992129.2 XP_013847583.1 559 63
XM_013992132.2 XP_013847586.1 564 63
GenBank mRNA AK390125.1 - 3 - 4 202 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

SNARE protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0