This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06903403_g1
See other VSIG2 GE Assays ›
Gene Symbol:
VSIG2
Gene Name:
V-set and immunoglobulin domain containing 2
Gene Aliases:
-
Chromosome Location:
Chr.9: 51974577 - 51982884 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06903403_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: VSIG2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021063059.1 XP_020918718.1 1661 103
XM_021063060.1 XP_020918719.1 1661 103
XM_021063061.1 XP_020918720.1 1502 103
GenBank mRNA AK232045.1 - 5 - 6 732 103
AK232048.1 - 5 - 6 800 103
AK232152.1 - 5 - 6 831 103
AK345778.1 - 5 - 6 796 103
AK345783.1 - 5 - 6 818 103
AK345786.1 - 5 - 6 748 103

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

defense/immunity protein immunoglobulin receptor superfamily

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0