Assay Details
Gene Symbol: SH3GL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003123066.4 | XP_003123114.1 | 1702 | 102 | ||
XM_005661329.2 | XP_005661386.1 | 1700 | 102 | |||
GenBank mRNA | AK231986.1 | - | 1 | 514 | 102 | |
AK236089.1 | - | 10 | 1679 | 102 | ||
AK393451.1 | - | 10 | 1703 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
