This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06903015_m1
See other CYRIA GE Assays ›
Gene Symbol:
CYRIA
Gene Name:
CYFIP related Rac1 interactor A
Gene Aliases:
FAM49A
Chromosome Location:
Chr.3: 120968016 - 121069372 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06903015_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CYRIA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003125345.5 XP_003125393.1 338 98
XM_005662738.3 XP_005662795.1 344 98
XM_013996336.2 XP_013851790.1 1801 98
GenBank mRNA AK231887.1 - 3 - 4 306 98
AK345847.1 - 3 - 4 300 98
AK390021.1 - 3 - 4 298 98
AK390341.1 - 3 - 4 302 98
AK400287.1 - 3 - 4 305 98

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0