This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06901712_m1
See other HDHD2 GE Assays ›
Gene Symbol:
HDHD2
Gene Name:
haloacid dehalogenase like hydrolase domain containing 2
Gene Aliases:
-
Chromosome Location:
Chr.1: 96682632 - 96729510 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06901712_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HDHD2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003122291.4 XP_003122339.1 779 56
XM_005652472.3 XP_005652529.1 821 56
XM_005652473.3 XP_005652530.1 925 56
XM_005652474.3 XP_005652531.1 800 56
XM_005652475.3 XP_005652532.1 784 56
XM_013978323.2 XP_013833777.1 978 56
XM_013978361.2 XP_013833815.1 771 56
XM_013978391.2 XP_013833845.1 913 56
XM_013978414.2 XP_013833868.1 807 56
XM_013978442.2 XP_013833896.1 643 56
XM_021092751.1 XP_020948410.1 858 56
XM_021092756.1 XP_020948415.1 1015 56
XM_021092764.1 XP_020948423.1 961 56
XM_021092772.1 XP_020948431.1 959 56
GenBank mRNA AK351846.1 - 6 - 7 898 56

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

phosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0