This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06900785_g1
See other CDKN1C GE Assays ›
Gene Symbol:
CDKN1C
Gene Name:
cyclin dependent kinase inhibitor 1C
Gene Aliases:
-
Chromosome Location:
Chr.2: 2019391 - 2022092 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06900785_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CDKN1C
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021082622.1 XP_020938281.1 1185 98
GenBank mRNA AK350718.1 - 3 - 4 951 98
AK395505.1 - 3 - 4 946 98
HQ679903.1 - 3 - 4 952 98

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

kinase inhibitor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0