This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06900589_m1
See other STYXL1 GE Assays ›
Gene Symbol:
STYXL1
Gene Name:
serine/threonine/tyrosine interacting like 1
Gene Aliases:
-
Chromosome Location:
Chr.3: 10131508 - 10212611 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06900589_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: STYXL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021086296.1 XP_020941955.1 1764 82
XM_021086297.1 XP_020941956.1 1161 82
XM_021086298.1 XP_020941957.1 1579 82
XM_021086299.1 XP_020941958.1 977 82
XM_021086300.1 XP_020941959.1 860 82
XM_021086301.1 XP_020941960.1 1659 82
XM_021086302.1 XP_020941961.1 1473 82
XM_021086303.1 XP_020941962.1 872 82
XM_021086304.1 XP_020941963.1 811 82
XM_021086305.1 XP_020941964.1 1013 82
XM_021086306.1 XP_020941965.1 1269 82
GenBank mRNA AK350484.1 - 9 - 10 1166 82
AK350980.1 - 7 - 8 1096 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0