Assay Details
Gene Symbol: CSRNP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003132123.4 | XP_003132171.1 | 715 | 113 | ||
GenBank mRNA | AK349562.1 | - | 3 - 4 | 695 | 113 | |
AK394156.1 | - | 3 - 4 | 671 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
