This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06898495_g1
See other CYBC1 GE Assays ›
Gene Symbol:
CYBC1
Gene Name:
cytochrome b-245 chaperone 1
Gene Aliases:
C12H17orf62
Chromosome Location:
Chr.12: 656776 - 662519 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06898495_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CYBC1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_013989815.2 XP_013845269.1 526 56
XM_013989817.2 XP_013845271.1 1209 56
XM_013989818.2 XP_013845272.1 655 56
XM_013989819.2 XP_013845273.1 685 56
XM_021066350.1 XP_020922009.1 1775 56
XM_021066351.1 XP_020922010.1 1872 56
XM_021066352.1 XP_020922011.1 864 56
XM_021066353.1 XP_020922012.1 774 56
GenBank mRNA AK349285.1 - 5 - 6 1003 56

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0