This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ss06898318_m1
See other LOC102162486 GE Assays ›
Gene Symbol:
LOC102162486
Gene Name:
putative RNA-binding protein Luc7-like 2
Gene Aliases:
LUC7L2
Chromosome Location:
Chr.18: 10268077 - 10333227 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID
Ss06898318_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: LOC102162486
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_005657737.3
XP_005657794.1
1247
159
XM_005657738.3
XP_005657795.1
1084
159
XM_005657742.3
XP_005657799.1
791
159
XM_005657744.3
XP_005657801.1
746
159
XM_013990775.2
XP_013846229.1
1315
159
XM_013990776.2
XP_013846230.1
3502
159
XM_013990777.2
XP_013846231.1
5159
159
XM_013990778.2
XP_013846232.1
4220
159
XM_013990779.2
XP_013846233.1
3424
159
XM_013990780.2
XP_013846234.1
1213
159
XM_013990781.2
XP_013846235.1
1163
159
XM_021078846.1
XP_020934505.1
3304
159
XM_021078847.1
XP_020934506.1
9415
159
XM_021078848.1
XP_020934507.1
3307
159
XM_021078849.1
XP_020934508.1
11073
159
XM_021078850.1
XP_020934509.1
2750
159
XM_021078851.1
XP_020934510.1
3231
159
XM_021078852.1
XP_020934511.1
3726
159
XM_021078853.1
XP_020934512.1
812
159
XM_021078854.1
XP_020934513.1
1093
159
XM_021078855.1
XP_020934514.1
3165
159
XM_021078856.1
XP_020934515.1
1369
159
XM_021078857.1
XP_020934516.1
1402
159
XM_021078858.1
XP_020934517.1
1240
159
GenBank mRNA
AK349205.1
-
6 - 7
1002
159
Back To Top
More Information
Set Membership:
Amplicon length greater than or equal to 101
3' Most
Sus scrofa
Probe spans exons
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header