This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06898317_m1
See other LOC102162486 GE Assays ›
Gene Symbol:
LOC102162486
Gene Name:
putative RNA-binding protein Luc7-like 2
Gene Aliases:
LUC7L2
Chromosome Location:
Chr.18: 10268077 - 10333227 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06898317_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC102162486
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005657737.3 XP_005657794.1 1072 90
XM_005657738.3 XP_005657795.1 909 90
XM_005657742.3 XP_005657799.1 616 90
XM_005657744.3 XP_005657801.1 571 90
XM_013990775.2 XP_013846229.1 1140 90
XM_013990776.2 XP_013846230.1 3327 90
XM_013990777.2 XP_013846231.1 4984 90
XM_013990778.2 XP_013846232.1 4045 90
XM_013990779.2 XP_013846233.1 3249 90
XM_013990780.2 XP_013846234.1 1038 90
XM_013990781.2 XP_013846235.1 988 90
XM_021078846.1 XP_020934505.1 3129 90
XM_021078847.1 XP_020934506.1 9240 90
XM_021078848.1 XP_020934507.1 3132 90
XM_021078849.1 XP_020934508.1 10898 90
XM_021078850.1 XP_020934509.1 2575 90
XM_021078851.1 XP_020934510.1 3056 90
XM_021078852.1 XP_020934511.1 3551 90
XM_021078853.1 XP_020934512.1 637 90
XM_021078854.1 XP_020934513.1 918 90
XM_021078855.1 XP_020934514.1 2990 90
XM_021078856.1 XP_020934515.1 1194 90
XM_021078857.1 XP_020934516.1 1227 90
XM_021078858.1 XP_020934517.1 1065 90
GenBank mRNA AK349205.1 - 5 - 6 827 90

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm