This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06895818_g1
See other LOC110260088 GE Assays ›
Gene Symbol:
LOC110260088
Gene Name:
eukaryotic translation initiation factor 3 subunit C
Gene Aliases:
-
Chromosome Location:
Chr.3: 18467725 - 18489088 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06895818_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC110260088
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021088039.1 XP_020943698.1 148 107
GenBank mRNA AK347191.1 - 1 - 2 75 107

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_021088039.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

translation initiation factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0