This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06895340_m1
See other PGPEP1 GE Assays ›
Gene Symbol:
PGPEP1
Gene Name:
pyroglutamyl-peptidase I
Gene Aliases:
-
Chromosome Location:
Chr.2: 59448110 - 59485809 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06895340_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PGPEP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021083522.1 XP_020939181.1 423 66
XM_021083523.1 XP_020939182.1 326 66
XM_021083524.1 XP_020939183.1 333 66
XM_021083525.1 XP_020939184.1 265 66
GenBank mRNA AK346443.1 - 4 - 5 358 66

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_021083525.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

cysteine protease protease protein modifying enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0