This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ss06894608_m1
See other HHLA2 GE Assays ›
Gene Symbol:
HHLA2
Gene Name:
HERV-H LTR-associating 2
Gene Aliases:
-
Chromosome Location:
Chr.13: 151153982 - 151298324 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID
Ss06894608_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
374.00
Your Price
Online offer:
374.0
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: HHLA2
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_005670241.3
XP_005670298.1
1419
102
XM_005670242.3
XP_005670299.1
1583
102
XM_005670245.3
XP_005670302.1
1513
102
XM_005670246.3
XP_005670303.1
1604
102
XM_013982469.2
XP_013837923.1
1665
102
XM_013982470.2
XP_013837924.1
1341
102
XM_013982471.2
XP_013837925.1
1944
102
XM_013982472.2
XP_013837926.1
1329
102
XM_013982473.2
XP_013837927.1
1299
102
XM_021070436.1
XP_020926095.1
1684
102
XM_021070437.1
XP_020926096.1
1301
102
XM_021070438.1
XP_020926097.1
1536
102
XM_021070439.1
XP_020926098.1
1435
102
XM_021070440.1
XP_020926099.1
1505
102
XM_021070441.1
XP_020926100.1
1467
102
XM_021070443.1
XP_020926102.1
2037
102
XM_021070444.1
XP_020926103.1
2107
102
XM_021070445.1
XP_020926104.1
2059
102
XM_021070446.1
XP_020926105.1
1211
102
XM_021070447.1
XP_020926106.1
1866
102
XM_021070448.1
XP_020926107.1
2030
102
XM_021070449.1
XP_020926108.1
1738
102
XM_021070451.1
XP_020926110.1
1251
102
XM_021070452.1
XP_020926111.1
1221
102
XM_021070453.1
XP_020926112.1
1824
102
XM_021070454.1
XP_020926113.1
1988
102
GenBank mRNA
AK345908.1
-
6 - 7
1203
102
Back To Top
More Information
Set Membership:
Amplicon length greater than or equal to 101
3' Most
Recommended Assay
Sus scrofa
Probe spans exons
Panther Classification:
Molecular Function -
immunoglobulin receptor superfamily
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0