Assay Details
Gene Symbol: ZNF577 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK345787.1 | - | 1 - 2 | 174 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |

Gene Symbol: ZNF577 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK345787.1 | - | 1 - 2 | 174 | 101 |
Set Membership: |
![]() ![]() ![]() |