Assay Details
Gene Symbol: CENPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003125331.6 | XP_003125379.1 | 541 | 60 | ||
XM_013996300.2 | XP_013851754.1 | 537 | 60 | |||
GenBank mRNA | AK344566.1 | - | 4 - 5 | 435 | 60 | |
AK389758.1 | - | 4 - 5 | 430 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
