This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06892387_m1
See other MTFR1L GE Assays ›
Gene Symbol:
MTFR1L
Gene Name:
mitochondrial fission regulator 1 like
Gene Aliases:
-
Chromosome Location:
Chr.6: 83265351 - 83277774 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06892387_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MTFR1L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003127710.4 XP_003127758.1 3202 78
XM_021095596.1 XP_020951255.1 492 78
XM_021095597.1 XP_020951256.1 484 78
XM_021095598.1 XP_020951257.1 483 78
XM_021095599.1 XP_020951258.1 485 78
GenBank mRNA AK344323.1 - 4 - 5 386 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0