Assay Details
Gene Symbol: CLEC2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003126492.4 | XP_003126540.1 | 1663 | 99 | ||
XM_003126493.4 | XP_003126541.1 | 1663 | 99 | |||
XM_021092371.1 | XP_020948030.1 | 1553 | 99 | |||
GenBank mRNA | AK343744.1 | - | 6 - 7 | 803 | 99 | |
AK343943.1 | - | 6 - 7 | 787 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
