Assay Details
Gene Symbol: RNPEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021064527.1 | XP_020920186.1 | 1051 | 58 | ||
GenBank mRNA | AK343500.1 | - | 2 - 3 | 282 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
