This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06888515_m1
See other RIPOR2 GE Assays ›
Gene Symbol:
RIPOR2
Gene Name:
RHO family interacting cell polarization regulator 2
Gene Aliases:
FAM65B
Chromosome Location:
Chr.7: 19609358 - 19842345 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06888515_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RIPOR2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005665637.3 XP_005665694.1 5307 76
XM_013977480.2 XP_013832934.1 7331 76
XM_013977481.2 XP_013832935.1 970 76
XM_013977485.2 XP_013832939.1 5307 76
XM_021098402.1 XP_020954061.1 1931 76
XM_021098404.1 XP_020954063.1 2727 76
XM_021098405.1 XP_020954064.1 7331 76
XM_021098406.1 XP_020954065.1 943 76
GenBank mRNA AK239122.1 - 3 - 4 260 76
AK398473.1 - 8 - 9 996 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0