This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06888507_m1
See other DARS1 GE Assays ›
Gene Symbol:
DARS1
Gene Name:
aspartyl-tRNA synthetase 1
Gene Aliases:
DARS
Chromosome Location:
Chr.15: 15810497 - 15885706 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06888507_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003133266.5 XP_003133314.1 686 101
XM_021075843.1 XP_020931502.1 1072 101
GenBank mRNA AK239116.1 - 6 - 7 616 101
AK390493.1 - 6 - 7 630 101

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

aminoacyl-tRNA synthetase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0