This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06888255_m1
See other TLE5 GE Assays ›
Gene Symbol:
TLE5
Gene Name:
TLE family member 5, transcriptional modulator
Gene Aliases:
AES
Chromosome Location:
Chr.2: 75492208 - 75501543 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06888255_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TLE5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021084158.1 XP_020939817.1 205 76
GenBank mRNA AK238961.1 - 1 - 2 180 76

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_021084158.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

transcription cofactor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0