This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06888112_m1
See other RORC GE Assays ›
Gene Symbol:
RORC
Gene Name:
RAR related orphan receptor C
Gene Aliases:
-
Chromosome Location:
Chr.4: 97374672 - 97399017 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06888112_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RORC
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003355171.4 XP_003355219.1 1104 51
XM_013997125.2 XP_013852579.1 1004 51
XM_013997126.2 XP_013852580.1 1004 51
XM_013997127.2 XP_013852581.1 1004 51
XM_013997128.2 XP_013852582.1 1004 51
XM_021089845.1 XP_020945504.1 1014 51
XM_021089846.1 XP_020945505.1 1014 51
XM_021089848.1 XP_020945507.1 1059 51
XM_021089849.1 XP_020945508.1 2298 51
XM_021089850.1 XP_020945509.1 2341 51
XM_021089851.1 XP_020945510.1 2302 51
XM_021089852.1 XP_020945511.1 1101 51
XM_021089853.1 XP_020945512.1 1872 51
GenBank mRNA AK238874.1 - 5 - 6 1063 51

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

C4 zinc finger nuclear receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0