This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06886709_m1
See other MIF4GD GE Assays ›
Gene Symbol:
MIF4GD
Gene Name:
MIF4G domain containing
Gene Aliases:
-
Chromosome Location:
Chr.12: 6047295 - 6052203 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06886709_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIF4GD
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003131204.4 XP_003131252.1 620 70
XM_005668610.3 XP_005668667.1 681 70
XM_013980750.2 XP_013836204.1 691 70
GenBank mRNA AK231263.1 - 5 - 6 622 70
AK232512.1 - 5 - 6 1262 70
AK237918.1 - 5 - 6 598 70
AK345638.1 - 5 - 6 584 70
AK392027.1 - 5 - 6 1217 70
AK400435.1 - 5 - 6 606 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0