This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06886312_m1
See other NDUFB4 GE Assays ›
Gene Symbol:
NDUFB4
Gene Name:
NADH:ubiquinone oxidoreductase subunit B4
Gene Aliases:
-
Chromosome Location:
Chr.13: 139724145 - 139730492 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06886312_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NDUFB4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003361882.4 XP_003361930.1 387 85
GenBank mRNA AK231094.1 - 2 - 3 341 85
AK231097.1 - 2 - 3 341 85
AK344760.1 - 2 - 3 389 85

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

oxidoreductase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm