This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06886243_m1
See other PGAP4 GE Assays ›
Gene Symbol:
PGAP4
Gene Name:
post-GPI attachment to proteins GalNAc transferase 4
Gene Aliases:
TMEM246
Chromosome Location:
Chr.1: 243167328 - 243190281 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06886243_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PGAP4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003353597.4 XP_003353645.1 235 56
GenBank mRNA AK231078.1 - 1 - 2 156 56

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_003353597.4 transcripts.

Set Membership:

Amplicon length less than or equal to 70 3' Most Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0