This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06885273_m1
See other MIA2 GE Assays ›
Gene Symbol:
MIA2
Gene Name:
MIA SH3 domain ER export factor 2
Gene Aliases:
CTAGE5
Chromosome Location:
Chr.1: 169777581 - 169868308 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06885273_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIA2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005659906.3 XP_005659963.1 2145 123
XM_005659907.3 XP_005659964.1 1525 123
XM_005659909.3 XP_005659966.1 2226 123
XM_013993367.2 XP_013848821.1 2117 123
XM_021073967.1 XP_020929626.1 3503 123
XM_021073969.1 XP_020929628.1 3488 123
XM_021073974.1 XP_020929633.1 3503 123
XM_021073975.1 XP_020929634.1 3476 123
XM_021074004.1 XP_020929663.1 1570 123
XM_021074012.1 XP_020929671.1 1658 123
XM_021074016.1 XP_020929675.1 1535 123
XM_021074027.1 XP_020929686.1 1664 123
XM_021074030.1 XP_020929689.1 1536 123
GenBank mRNA AK237997.1 - 6 - 7 503 123

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

membrane traffic protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0