Assay Details
Gene Symbol: MIA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005659906.3 | XP_005659963.1 | 2038 | 94 | ||
XM_005659907.3 | XP_005659964.1 | 1418 | 94 | |||
XM_005659909.3 | XP_005659966.1 | 2119 | 94 | |||
XM_013993367.2 | XP_013848821.1 | 2010 | 94 | |||
XM_021073967.1 | XP_020929626.1 | 3396 | 94 | |||
XM_021073969.1 | XP_020929628.1 | 3381 | 94 | |||
XM_021073974.1 | XP_020929633.1 | 3396 | 94 | |||
XM_021073975.1 | XP_020929634.1 | 3369 | 94 | |||
XM_021074004.1 | XP_020929663.1 | 1463 | 94 | |||
XM_021074012.1 | XP_020929671.1 | 1551 | 94 | |||
XM_021074016.1 | XP_020929675.1 | 1428 | 94 | |||
XM_021074027.1 | XP_020929686.1 | 1557 | 94 | |||
XM_021074030.1 | XP_020929689.1 | 1429 | 94 | |||
GenBank mRNA | AK237997.1 | - | 4 - 5 | 396 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
