This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06883904_m1
See other BCAP31 GE Assays ›
Gene Symbol:
BCAP31
Gene Name:
B cell receptor associated protein 31
Gene Aliases:
-
Chromosome Location:
Chr.X: 124456999 - 124484923 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06883904_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BCAP31
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003135480.4 XP_003135528.2 660 64
XM_005674003.2 XP_005674060.1 629 64
XM_005674004.2 XP_005674061.1 517 64
XM_021079672.1 XP_020935331.1 629 64
GenBank mRNA AK237142.1 - 4 - 5 454 64
AK348920.1 - 3 - 4 398 64
AK351227.1 - 4 - 5 440 64
AK389367.1 - 3 - 4 628 64
AK391722.1 - 4 - 5 430 64
AK398144.1 - 4 - 5 481 64
AK398229.1 - 4 - 5 509 64
AK400474.1 - 4 - 5 644 64
AK400971.1 - 4 - 5 577 64
AY609758.1 - 4 - 5 558 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

membrane traffic protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0