Assay Details
Gene Symbol: DDX19B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003126872.4 | XP_003126920.1 | 633 | 73 | ||
XM_005653242.3 | XP_005653299.1 | 727 | 73 | |||
GenBank mRNA | AK237089.1 | - | 6 - 7 | 621 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
