This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06883741_g1
See other NFKB2 GE Assays ›
Gene Symbol:
NFKB2
Gene Name:
nuclear factor kappa B subunit 2
Gene Aliases:
NFKB2_TV2
Chromosome Location:
Chr.14: 113406412 - 113414177 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06883741_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NFKB2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021072741.1 XP_020928400.1 59
XM_021072742.1 XP_020928401.1 59
XM_021072743.1 XP_020928402.1 59
XM_021072744.1 XP_020928403.1 59
XM_021072745.1 XP_020928404.1 59
XM_021072746.1 XP_020928405.1 59
GenBank mRNA AK237079.1 - 1 - 2 112 59
AK396815.1 - 13 - 14 59

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Inventoried Probe spans exons

Panther Classification:

Molecular Function -

Rel homology transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0