This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06881820_m1
See other LOC100157433 GE Assays ›
Gene Symbol:
LOC100157433
Gene Name:
calcium homeostasis modulator protein 2
Gene Aliases:
CALHM1, CALHM2
Chromosome Location:
Chr.14: 114380467 - 114393845 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06881820_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100157433
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021073485.1 XP_020929144.1 1976 96
XM_021073486.1 XP_020929145.1 1304 96
XM_021073487.1 XP_020929146.1 1363 96
XM_021073488.1 XP_020929147.1 1346 96
XM_021073489.1 XP_020929148.1 1270 96
GenBank mRNA AK236120.1 - 3 - 4 969 96
AK348440.1 - 2 - 3 994 96

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0