This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06881439_m1
See other NUDT6 GE Assays ›
Gene Symbol:
NUDT6
Gene Name:
nudix hydrolase 6
Gene Aliases:
-
Chromosome Location:
Chr.8: 101241155 - 101302760 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06881439_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NUDT6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003129210.4 XP_003129258.1 362 90
XM_005666886.3 XP_005666943.2 835 90
XM_005666888.3 XP_005666945.1 602 90
XM_005666889.3 XP_005666946.1 445 90
XM_005666890.3 XP_005666947.1 499 90
XM_013978915.2 XP_013834369.2 429 90
XM_013978916.2 XP_013834370.1 528 90
GenBank mRNA AK235922.1 - 4 - 5 615 90
AK352172.1 - 4 - 5 559 90
CK468379.1 - 4 - 5 477 90

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

nucleotide phosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0