This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06881047_m1
See other EMR4 GE Assays ›
Gene Symbol:
EMR4
Gene Name:
EGF-like module containing, mucin-like, hormone receptor-like sequence 4
Gene Aliases:
EMR4P
Chromosome Location:
Chr.2: 72155534 - 72206486 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06881047_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EMR4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021083944.1 XP_020939603.1 389 92
XM_021083945.1 XP_020939604.1 340 92
XM_021083946.1 XP_020939605.1 390 92
XM_021083947.1 XP_020939606.1 390 92
XM_021083948.1 XP_020939607.1 390 92
XM_021083949.1 XP_020939608.1 390 92
XM_021083950.1 XP_020939609.1 390 92
GenBank mRNA AK230928.1 - 4 - 5 314 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0