This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06880613_m1
See other PRDX1 GE Assays ›
Gene Symbol:
PRDX1
Gene Name:
peroxiredoxin 1
Gene Aliases:
-
Chromosome Location:
Chr.6: 165847390 - 165859918 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06880613_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRDX1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021096742.1 XP_020952401.1 1060 60
XM_021096743.1 XP_020952402.1 635 60
XM_021096744.1 XP_020952403.1 897 60
GenBank mRNA AK230806.1 - 8 - 9 965 60
AK231390.1 - 5 - 6 569 60
AK231993.1 - 5 - 6 600 60
AK346969.1 - 5 - 6 576 60
AK352082.1 - 5 - 6 571 60
AK392161.1 - 5 - 6 572 60
AY610415.1 - 5 - 6 542 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa Inventoried Probe spans exons

Panther Classification:

Molecular Function -

peroxidase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0