This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06880482_m1
See other PPIG GE Assays ›
Gene Symbol:
PPIG
Gene Name:
peptidylprolyl isomerase G
Gene Aliases:
-
Chromosome Location:
Chr.15: 75940397 - 75979168 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06880482_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPIG
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001926861.6 XP_001926896.1 349 72
XM_005671914.3 XP_005671971.1 519 72
XM_005671915.3 XP_005671972.1 397 72
XM_005671916.3 XP_005671973.1 569 72
XM_021076268.1 XP_020931927.1 576 72
XM_021076269.1 XP_020931928.1 381 72
XM_021076270.1 XP_020931929.1 552 72
XM_021076271.1 XP_020931930.1 429 72
GenBank mRNA AK230754.1 - 4 - 5 345 72
AK395757.1 - 4 - 5 348 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0