This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06880432_m1
See other RWDD2A GE Assays ›
Gene Symbol:
RWDD2A
Gene Name:
RWD domain containing 2A
Gene Aliases:
-
Chromosome Location:
Chr.1: 82997984 - 83003583 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06880432_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RWDD2A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001926568.3 XP_001926603.1 330 92
XM_013992798.2 XP_013848252.1 460 92
GenBank mRNA AK230738.1 - 2 - 3 401 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0