Assay Details
Gene Symbol: PKM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001929069.5 | XP_001929104.1 | 58 | |||
XM_005666187.3 | XP_005666244.1 | 58 | ||||
XM_005666188.3 | XP_005666245.1 | 58 | ||||
XM_013978108.2 | XP_013833562.1 | 58 | ||||
XM_021099106.1 | XP_020954765.1 | 58 | ||||
XM_021099107.1 | XP_020954766.1 | 58 | ||||
XM_021099118.1 | XP_020954777.1 | 58 | ||||
XM_021099119.1 | XP_020954778.1 | 58 | ||||
XM_021099121.1 | XP_020954780.1 | 58 | ||||
XM_021099122.1 | XP_020954781.1 | 58 | ||||
XM_021099123.1 | XP_020954782.1 | 58 | ||||
XM_021099124.1 | XP_020954783.1 | 58 | ||||
XM_021099125.1 | XP_020954784.1 | 58 | ||||
GenBank mRNA | AK235447.1 | - | 4 - 5 | 482 | 58 | |
AK235858.1 | - | 4 - 5 | 489 | 58 | ||
AK239164.1 | - | 2 - 3 | 58 | |||
AK390609.1 | - | 5 - 6 | 58 | |||
AK392642.1 | - | 4 - 5 | 58 | |||
AK394600.1 | - | 4 - 5 | 58 | |||
AK399587.1 | - | 4 - 5 | 58 | |||
AK399984.1 | - | 4 - 5 | 58 | |||
AK400779.1 | - | 4 - 5 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
