This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06877726_m1
See other GOLGA7 GE Assays ›
Gene Symbol:
GOLGA7
Gene Name:
golgin A7
Gene Aliases:
-
Chromosome Location:
Chr.17: 10624378 - 10643844 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06877726_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GOLGA7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003134222.5 XP_003134270.1 200 78
XM_005672664.3 XP_005672721.1 155 78
XM_005672665.3 XP_005672722.1 200 78
XM_021077541.1 XP_020933200.1 200 78
XM_021077542.1 XP_020933201.1 202 78
XM_021077543.1 XP_020933202.1 156 78
XM_021077545.1 XP_020933204.1 200 78
GenBank mRNA AK234963.1 - 2 - 3 148 78
AK399258.1 - 2 - 3 170 78
AY609462.1 - 2 - 3 183 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0