Assay Details
Gene Symbol: GDPGP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003482236.4 | XP_003482284.1 | 126 | 107 | ||
GenBank mRNA | AK234600.1 | - | 1 - 2 | 75 | 107 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003482236.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
