This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06876749_g1
See other RPS14 GE Assays ›
Gene Symbol:
RPS14
Gene Name:
ribosomal protein S14
Gene Aliases:
-
Chromosome Location:
Chr.2: 151430047 - 151433951 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06876749_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPS14
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021085026.1 XP_020940685.1 525 80
XM_021085027.1 XP_020940686.1 525 80
XM_021085028.1 XP_020940687.1 527 80
GenBank mRNA AK234565.1 - 4 - 5 447 80
AK238512.1 - 4 - 5 425 80
AK345144.1 - 4 - 5 424 80
AK346810.1 - 4 - 5 524 80
AK347086.1 - 4 - 5 445 80
AK347408.1 - 4 - 5 532 80
AK347495.1 - 4 - 5 445 80
AK348868.1 - 4 - 5 445 80
AK350474.1 - 4 - 5 467 80
AK351156.1 - 4 - 5 448 80
AK351311.1 - 4 - 5 449 80
AK392020.1 - 4 - 5 450 80
AK397603.1 - 4 - 5 422 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

ribosomal protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0